VWR Healthcare Clinical Catalog 2012-2014 - page 356
Eppendorf
®
Mastercycler
®
pro and pro S Thermal Cyclers
Thermal cyclers feature heated lids with vapo.protect
technology to reduce evaporation to a minimum and a freely program-
mable temperature gradient over 12 rows. Triple circuit technology ensures a homogeneous temperature distibution and
enables the generation of specific temperature gradients for PCR optimizations. SteadySlope
®
technology ensures that the
heating and cooling ramp rates for thermal modules are identical in both gradient and normal operation, and TSP (Thermal
Sample Protection) technology reduces the thermal load on the samples and minimizes the probability of non-specific product
formation during PCR. Expandable to a mini satellite system or network.
The Mastercycler
®
pro cycler features a robust aluminum block and a gradient range of up to 20°C (36°F).
The Mastercycler
®
pro S cycler features a silver block that provides the highest temperature control speed and extremely fast
heating and cooling rates, a pulse function for device-driven hot start for avoiding non-specific PCR product formation, and an
extended gradient range of up to 24°C (43.2°F).
Ordering Information:
A control panel or CycleManager pro software (both sold separately) is required for operation. Both
the control panel and CycleManager pro software include a connection cable. CAN-Bus connection cables (not included) are
required to link cyclers in an network. For CAN-Bus connection cables, service programs, and other accessories, contact your
VWR representative.
Model
Mastercycler
®
pro
Mastercycler
®
pro S
Block homogeneity (°C)
≤ ±0.3°C at 20–72°C (68–161.6°F), ≤ ±0.4°C at 95°C (203°F)
≤ ±0.3°C at 20–72°C (68–161.6°F), ≤ ±0.4°C at 95°C (203°F)
Block Temperature Range
4–99°C (39.2–210.2°F)
4–99°C (39.2–210.2°F)
Control Accuracy
±0.2°C (±0.36°F)
±0.2°C (±0.36°F)
Cooling Rate
3°C/s (5.4°F/s), measured at block
4.5°C/s (8.1°F/s), measured at block
Electrical
120V, 50-60Hz, 950W
120V, 50Hz, 950W
Gradient Range
1–20°C (1.8–36°F)
1–24°C (1.8–43.2°F)
Gradient temperature range (°C)
30–99°C (86–210.2°F)
30–99°C (86–210.2°F)
Heating Rate
4°C/s (7.2°F/s), measured at block
6°C/s (10.8°F/s), measured at block
Lid temperature range (°C)
37–110°C (98.6–230°F)
37–110°C (98.6–230°F)
Sample capacity
96 x 0.2 mL PCR tubes or a 96-well PCR plate
96 x 0.2 mL PCR tubes or a 96-well PCR plate
Weight
18.5 kg (40.8 lbs.)
18.5 kg (40.8 lbs.)
W x D x H
26 × 41.5 × 37 cm (10
1
/
4
x 16
11
/
32
x 14
9
/
16
")
26 × 41.5 × 37 cm (10
1
/
4
x 16
11
/
32
x 14
9
/
16
")
Description
Supplier No.
Cat. No.
Price
Mastercycler
®
pro Thermal Cycler
950030010
Ea./
7828.00
Mastercycler
®
pro S Thermal Cycler
950030020
Ea./
8697.00
CycleManager pro Software
950017007
Ea./
2278.00
Control Panel
950030050
Ea./
2164.00
1.800.932.5000
|
vwr.com
|
VWR International
|
343
Molecular Diagnostics
Thermal Cyclers
1...,346,347,348,349,350,351,352,353,354,355 357,358,359,360,361,362,363,364,365,366,...955
Powered by FlippingBook