VWR Healthcare Clinical Catalog 2012-2014 - page 355
TC-5000 Series PCR Gradient Thermal Cyclers, Techne
®
TC-5000 series thermal cyclers feature an intuitive touch screen interface, real-time graphical display, gradient capabilities, gradi-
ent calculator, and a user-adjustable heated lid. Include independent over-temperature cutout and power loss auto restart functions.
Formats include a 96x0.2mL, 60x0.5mL, 384-well, or in situ block.
Ordering Information:
Four-year manufacturer's warranty. The 384-well block, fully skirted plate, and in situ block are special order
block formats; contact your VWR representative for more information.
Block Temperature Range
4 to 99°C
Block Uniformity at 50°C
±0.3°C
Gradient Range
20 to 70°C
Maximum Gradient
30°C (16°C for 384-well block) (unavailable for in situ block)
Maximum Ramp Rate
3.0°C/sec.
No. of Programs
50
Peltier Elements per
Block
8
Block Format
Supplier No.
Cat. No.
Price
96 x 0.2 mL
FTC5/H02
Ea./
6860.00
96 x 0.2 mL Fully Skirted Plate
FTC5/F02
Ea./
6860.00
60 x 0.5 mL
FTC5/05
Ea./
6860.00
384-Well Plate
FTC5/384
Ea./
6860.00
Flat Plate For In situ
FTC5/FLAT
Ea./
6860.00
Eppendorf
®
Mastercycler
®
Thermal Cyclers
PCR-authorized thermal cyclers feature Peltier technology for heating and cooling, temperature control from 4 to 99°C (39 to 210°F),
simple programming, and LCD readout. Cyclers feature RS-232 and printer interface connections. Personal memory card can store up
to 10 individually-developed protocols, allowing easy data transfer between Mastercycler models. Choose from Mastercycler Gradient,
Mastercycler Personal, or Mastercycler thermal cyclers.
Ordering Information:
Manufacturer’s two-year warranty. Thermal cyclers include a heated lid and one personal memory card. Order
additional personal memory cards separately.
Control Accuracy
±0.2°C (0.4°F)
Cooling Control Speed
Approx. 2°C/s (3.6°F)
Heating Control Speed
Approx. 3°C/s (5.4°F)
Maximum Number of Cycles
99
Temperature Uniformity Across the Block
95°C (203°F), ±1.0°C (1.8°F), 20 to 72°C (68 to 161.6°F), ±0.6°C (1.1°F)
Description
Supplier No.
Cat. No.
Price
Mastercycler, 115V, 60Hz
950000058
Ea./
7284.00
Mastercycler Gradient, 115V, 60Hz
950000015
Ea./
7844.00
Mastercycler Gradient, 220V, 50Hz
950000023
Ea./
7844.00
Mastercycler Personal, 115V, 60Hz
950000031
Ea./
4479.00
Mastercycler Personal, 220V, 50Hz
950000040
Ea./
4479.00
342
|
VWR International
|
vwr.com
|
1.800.932.5000
Molecular Diagnostics
Thermal Cyclers
1...,345,346,347,348,349,350,351,352,353,354 356,357,358,359,360,361,362,363,364,365,...955
Powered by FlippingBook