BDH Chemicals 2013 - page 74
SODIUM HYDROXIDE/THIOSL 50%
Sodium hydroxide 50.0% - sodium
thiosulfate solution 2.5%
Made with deionized water.
Appearance/color................................................................................................................................Clear
Shelf-life........................................................................................................................................... 1 year
Size
Packaging
Cat. No.
Price
4 L
Poly Bottle
Ea./
50.44
Cs. 4/
206.77
SODIUM HYDROXIDE, 50%
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1.53 g/cm³ (20°C)
CAS:
1310-73-2
Sodium Hydroxide, Solution. 50% (w/w).
Made with deionized water.
Sodium hydroxide.....................................................................................................................50-52% w/w
Sodium carbonate.............................................................................................................. 0.00-0.25% w/w
Chloride........................................................................................................................0.000-0.005 % w/w
Heavy metals (as Ag) by EPA-6010B.......................................................................................... 0.0005% w/w
Iron (Fe) by EPA-6010B............................................................................................................ 0.0005% w/w
Nickel (Ni)............................................................................................................................. 0.0005% w/w
Nirtogen compounds (as N)..................................................................................................... 0.0005% w/w
Phosphate...............................................................................................................................0.025% w/w
Potassium................................................................................................................................0.003% w/w
Size
Packaging
Cat. No.
Price
4 L
Poly Bottle
Ea./
74.28
Cs. 4/
296.08
Sodium Hydroxide, Solution. Reagent Grade. 50% (w/w).
Clear liquid. Corrosive and toxic.
APHA color.....................................................................................................................................20 max.
Shelf-life..........................................................................................................................................3 years
Sodium hydroxide..................................................................................................................50-52 % (w/w)
Sodium carbonate........................................................................................................... 0.00-0.25 % (w/w)
Chloride................................................................................................................................ 0.000-0.005%
Heavy metals (as Ag) by EPA-6010B..................................................................................... 0.002 % max. wt.
Iron (Fe) by 6010B............................................................................................................0.0005 % wt. max.
Nickel (Ni).............................................................................................................................0.0005% max.
Nitrogen compounds (as N).....................................................................................................0.0005% max.
Phosphate.............................................................................................................................0.0005% max.
Potassium................................................................................................................................0.025% max.
Sulfate (SO
4
)............................................................................................................................0.003% max.
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
27.28
Cs. 12/
311.04
4 L
Poly Bottle
Ea./
55.63
Cs. 4/
227.99
10 L
Poly Drum
Ea./
92.73
20 L
NAMPAC®
Ea./
167.95
SODIUM HYDROXIDE 33%
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
CAS:
1310-73-2
Sodium hydroxide 33% (w/w) solution
Sodium hydroxide 33% (w/w) in purified water.
Appearance/color................................................................................................................................Clear
Sodium hydroxide............................................................................................................. 32.6-33.4% (w/w)
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
4 L
Poly Bottle
Ea./
36.57
Cs. 4/
146.27
EINECS:
215-185-5
ADR:
8,II
UN:
1824
EINECS:
215-185-5
ADR:
8,II
UN:
1824
SODIUM HYDROXIDE
Caustic soda
Formula:
NaOH
MW:
40 g/mol
Boiling Pt:
1390°C (1013 hPa)
Melting Pt:
323°C
Density:
2.13 g/cm³ (20°C)
Sodium Hydroxide, ACS Grade.
Beads.
Assay........................................................................................................................................... 97% min
Sodium Cabonate (Na2CO
3
)...........................................................................................................1.0% max
Sulfate (SO
4
)............................................................................................................................ 0.003% max
Chloride (Cl)............................................................................................................................. 0.005% max
Nitrogen compounds (as N)....................................................................................................... 0.001% max
Phosphate (PO
4
)....................................................................................................................... 0.001% max
Heavy metals (as Ag)................................................................................................................ 0.002% max
Iron (Fe).................................................................................................................................. 0.001% max
Nickel (Ni)............................................................................................................................... 0.001% max
Mercury (Hg)........................................................................................................................... 0.1 ppm max
Calcium (Ca)............................................................................................................................ 0.005% max
Magnesium (Mg)...................................................................................................................... 0.002% max
Potassium (K)............................................................................................................................. 0.02% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
18.00
Cs. 12/
141.55
2.5 kg
Poly Bottle
Ea./
108.86
Cs. 4/
380.94
12 kg
Poly Pail
Ea./
150.42
Pl. 27/
5734.41
CAS:
1310-73-2
EINECS:
215-185-5
ADR:
8,II
Index:
011-002-00-6
UN:
1823
NaOH
NaOH
NaOH
Get immediate answers to
your technical questions
If you have any questions about
chemicals and can’t wait for an answer,
contact our Technical Product Support
Team by calling
888.897.5463
, Monday-
Friday, 8
:
30am – 8
:
00pm, or email us at
.
A chemical expert will answer your call
and provide you with a real-time response
to your question or problem.
56
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
S
Sodium Hydroxide
1...,64,65,66,67,68,69,70,71,72,73 75,76,77,78,79,80,81,82,83,84,...100
Powered by FlippingBook