BDH Chemicals 2013 - page 72
SODIUM BISULFITE
Sodium Bisulfite, ACS
White solid
Assay................................................................................................................................... 58.5% SO
2
min
Insoluble matter....................................................................................................................... 0.005% max
Chloride (Cl)............................................................................................................................... 0.02% max
Heavy metals (as Pb)................................................................................................................. 0.001% max
Iron (Fe).................................................................................................................................. 0.002% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
39.36
Cs. 12/
333.70
SODIUM CARBONATE
Formula:
Na
2
CO
3
MW:
105.99 g/mol
Boiling Pt:
1600°C (1013 hPa)
Melting Pt:
854°C
Sodium Carbonate Anhydrous, ACS Grade.
White solid
Assay (dried basis)........................................................................................................... 99.5% Na2CO
3
min
Insoluble matter......................................................................................................................... 0.01% max
Loss on heating at 285°C...............................................................................................................1.0% max
Chloride (Cl)............................................................................................................................. 0.001% max
Phosphate (PO
4
)....................................................................................................................... 0.001% max
Silica (SiO
2
).................................................................................................................................... 0.00005
Sulfur compounds (as SO
4
)......................................................................................................... 0.003% max
Heavy metals (as Pb)................................................................................................................... 5 ppm max
Iron (Fe).................................................................................................................................... 5 ppm max
Calcium (Ca).............................................................................................................................. 0.03% max
Magnesium (Mg)...................................................................................................................... 0.005% max
Potassium (K)........................................................................................................................... 0.005% max
Size
Packaging Cat. No.
Price
500 g
Poly Bottle
Ea./
39.36
Cs. 12/
333.70
2.5 kg
Poly Bottle
Ea./
101.19
Cs. 4/
353.84
SODIUM CARBONATE 0.5 MOL/L (1 N)
AQUEOUS SOLUTION
Formula:
CNa
2
O
3
MW:
105.99 g/mol
Sodium Carbonate, Solution. 1N.
Made with deionized water. Suitable as a titrant.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.9995-1.0005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
34.21
Cs. 12/
389.95
CAS:
497-19-8
EINECS:
207-838-8
SODIUM CHLORIDE
Formula:
NaCl
MW:
58.44 g/mol
Boiling Pt:
1413°C (1013 hPa)
Melting Pt:
801°C
Sodium Chloride, ACS
White solid
Assay.................................................................................................................................... 99% NaCl min
pH of a solution at 25.0°C................................................................................................................5.0 – 9.0
Insoluble matter....................................................................................................................... 0.005% max
Iodide (I)................................................................................................................................. 0.002% max
Bromide (Br).............................................................................................................................. 0.01% max
Chlorate and nitrate (as NO
3
)..................................................................................................... 0.003% max
Phosphate (PO
4
)......................................................................................................................... 5 ppm max
Sulfate (SO
4
)............................................................................................................................ 0.004% max
Barium (Ba)................................................................................................................................Passes test
Heavy metals (as Pb)................................................................................................................... 5 ppm max
Iron (Fe).................................................................................................................................... 2 ppm max
Calcium (Ca)............................................................................................................................ 0.002% max
Magnesium (Mg)...................................................................................................................... 0.001% max
Potassium (K)........................................................................................................................... 0.005% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
31.43
Cs. 12/
265.21
2.5 kg
Poly Bottle
Ea./
64.74
Cs. 4/
225.89
12 kg
Poly Pail
Ea./
129.93
Pl. 27/
3069.23
250 lbs.
Fiber Drum
Ea./
473.40
Pl. 4/
1656.72
NaO ONa
O
NaO ONa
O
HO ONa
S
O
Density:
2.53 g/cm³ (20°C)
CAS:
497-19-8
EINECS:
207-838-8
Index:
011-005-00-2
Density:
2.16 g/cm³ (20°C)
CAS:
7647-14-5
EINECS:
231-598-3
NaCl
VWR has what you need for safe chemical handling and storage
VWR is your best source for complete personal protection in the lab. We also have comprehensive
chemical storage, disposal, and spill containment solutions. Connect with your VWR Sales
Representative or visit
us.vwr.com/safetyandindustrial
for more information today.
54
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
S
Sodium Bisulfite
1...,62,63,64,65,66,67,68,69,70,71 73,74,75,76,77,78,79,80,81,82,...100
Powered by FlippingBook