BDH Chemicals 2013 - page 70
QUARTZ
Silicium (IV) oxide, Silicium dioxide, Silicon dioxide, Sand, White quartz
Formula:
SiO₂
MW:
60.08 g/mol
Boiling Pt:
2230°C (1013 hPa)
Melting Pt:
1427°C
Density:
2.622 to 2.645 g/cm³ (20°C)
CAS:
14808-60-7
EINECS:
238-878-4
Sand, Ottawa
Beige granules
Moisture content..........................................................................................................................0.1% max
Particle size 30-40 mesh................................................................................................................. 80% min
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
29.43
Cs. 12/
230.26
2.5 kg
Poly Bottle
Ea./
40.97
Cs. 4/
114.91
SILVER NITRATE
Formula:
AgNO
3
MW:
169.87 g/mol
Boiling Pt:
444°C (1013 hPa)
Melting Pt:
210°C
Density:
4.35 g/cm³ (20°C)
Silver Nitrate, ACS
Colorless or white solid
Assay................................................................................................................................. 99% AgNO
3
min
Clarity of solution........................................................................................................................Passes test
Chloride (Cl)............................................................................................................................... 5 ppm max
Free acid....................................................................................................................................Passes test
Substances not precipitated by hydrochloric acid............................................................................ 0.01% max
Sulfate (SO
4
)............................................................................................................................ 0.002% max
Copper (Cu)................................................................................................................................ 2 ppm max
Iron (Fe).................................................................................................................................... 2 ppm max
Lead (Pb)................................................................................................................................. 0.001% max
Size
Packaging
Cat. No.
Price
125 g
Glass Bottle
Ea./
1064.03
Cs. 12/
11204.11
500 g
Glass Bottle
Ea./
4215.78
Cs. 12/
42221.95
SILVER NITRATE 0.5 MOL/L (0.5 N)
AQUEOUS SOLUTION
Formula:
AgNO
3
MW:
169.87 g/mol
CAS:
7761-88-8
Silver Nitrate Solution. 0.5N.
Made with deionized water.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.4995-0.5005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
Poly Bottle
Ea./
404.95
SILVER NITRATE 0.171 MOL/L (0.171
N, 29.05 G/L, 1 ML = 10 MG NACL)
AQUEOUS SOLUTION
Formula:
AgNO
3
MW:
169.87 g/mol
CAS:
7761-88-8
Silver Nitrate, Solution, 0.171N
Made with deionized water.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.1705-0.1715
Shelf-life.....................................................................................................................................18 months
Size
Packaging
Cat. No.
Price
4 L
Poly Bottle
Ea./
735.77
Cs. 4/
2769.94
CAS:
7761-88-8
EINECS:
231-853-9
ADR:
5.1,II
Index:
047-001-00-2
UN:
1493
EINECS:
231-853-9
ADR:
8,II
UN:
1760
EINECS:
231-853-9
ADR:
8,II
UN:
1760
SILVER NITRATE 0.1 MOL/L (0.1 N)
AQUEOUS SOLUTION
Formula:
AgNO
3
MW:
169.87 g/mol
Density:
1.015 g/cm³ (25°C)
Silver Nitrate, Solution. 0.1N.
Suitable as a titrant. Made with deionized water.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.0995-0.1005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
500 mL
Glass Bottle
Ea./
82.45
Cs. 12/
939.95
1 L
Poly Bottle
Ea./
200.89
Cs. 12/
2290.15
4 L
Amber Glass Bottle
Ea./
820.60
Cs. 4/
3254.09
4 L
Glass Bottle
Ea./
801.94
Cs. 4/
3207.77
20 L
Cubitainer®
Ea./
1710.28
CAS:
7761-88-8
EINECS:
231-853-9
SiO
2
AgNO
3
AgNO
3
AgNO
3
AgNO
3
Use the advanced search features
on VWR.com to find exactly what
you need fast
Now you can search by
CAS number, formula,
molecular weight, Merck
index, synonyms, or basic
chemical structure to
find the best chemical for
your needs.
52
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
Q
Quartz
1...,60,61,62,63,64,65,66,67,68,69 71,72,73,74,75,76,77,78,79,80,...100
Powered by FlippingBook