BDH Chemicals 2013 - page 65
POTASSIUM HYDROXIDE
Caustic potash
Formula:
KOH
MW:
56.11 g/mol
Boiling Pt:
1320°C (1013 hPa)
Melting Pt:
360°C
Density:
2.04 g/cm³ (20°C)
Potassium Hydroxide, ACS Grade.
White, deliquescent solid
Assay...........................................................................................................85.0% KOH min and 2.0% K
2
CO
3
Nitrogen compounds (as N)....................................................................................................... 0.001% max
Chloride (Cl)............................................................................................................................... 0.01% max
Phosphate (PO
4
)......................................................................................................................... 5 ppm max
Sulfate (SO
4
)............................................................................................................................ 0.003% max
Heavy metals (as Ag)................................................................................................................ 0.001% max
Iron (Fe).................................................................................................................................. 0.001% max
Nickel (Ni)............................................................................................................................... 0.001% max
Calcium (Ca)............................................................................................................................ 0.005% max
Magnesium (Mg)...................................................................................................................... 0.002% max
Sodium (Na)............................................................................................................................... 0.05% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
33.50
Cs. 12/
283.57
2.5 kg
Poly Bottle
Ea./
90.45
Cs. 4/
271.85
POTASSIUM HYDROXIDE
50% AQUEOUS SOLUTION
Formula:
KOH
MW:
56.11 g/mol
CAS:
1310-58-3
Potassium Hydroxide, Solution. 50% (w/v).
APHA for anaerobic sludge.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Potassium hydroxide % (w/v)...............................................................................................................48-52
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
48.75
Cs. 12/
555.75
POTASSIUM HYDROXIDE
45% AQUEOUS SOLUTION
Formula:
KOH
MW:
56.11 g/mol
CAS:
1310-58-3
CAS:
1310-58-3
EINECS:
215-181-3
ADR:
8,II
Index:
019-002-00-8
UN:
1813
EINECS:
215-181-3
ADR:
8,II
UN:
1814
EINECS:
215-181-3
ADR:
8,II
UN:
1814
Potassium Hydroxide, Solution. Technical Grade. 45% (w/w).
Shelf-life..........................................................................................................................................2 years
KOH............................................................................................................................................... 43-47%
K
2
CO
3
...................................................................................................................................... 0.10% max..
KCl......................................................................................................................................... 30 ppm max.
Iron (Fe) by EPA-6010B.............................................................................................................0.5 ppm max.
Heavy metals (as Pb).................................................................................................................. 5 ppm max.
Sodium...............................................................................................................................2500 ppm max.
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
51.15
Cs. 12/
583.10
4 L
Poly Bottle
Ea./
141.73
Cs. 4/
564.66
POTASSIUM HYDROXIDE
10% AQUEOUS SOLUTION
Formula:
KOH
MW:
56.11 g/mol
CAS:
1310-58-3
Potassium Hydroxide, Solution. 10% (w/v).
Made with deionized water.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Shelf-life..........................................................................................................................................2 years
Potassium hydroxide % (w/v)........................................................................................................... 9.9-10.1
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
35.02
Cs. 12/
399.23
EINECS:
215-181-3
ADR:
8,II
UN:
1814
POTASSIUM HYDROXIDE 1 MOL/L
(1 N) AQUEOUS SOLUTION
Formula:
KOH
MW:
56.11 g/mol
Density:
1.05 g/cm³ (25°C)
CAS:
1310-58-3
Potassium Hydroxide, Solution. 1.0N.
Made with deionized water. Suitable as a titrant.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.9995-1.0005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
31.56
Cs. 12/
359.77
EINECS:
215-181-3
ADR:
8,II
UN:
1814
800.932.5000
|
vwr.com
|
VWR International
|
47
P
Potassium Hydroxide
KOH
KOH
KOH
KOH
KOH
1...,55,56,57,58,59,60,61,62,63,64 66,67,68,69,70,71,72,73,74,75,...100
Powered by FlippingBook