BDH Chemicals 2013 - page 64
POTASSIUM CHLORIDE 3 MOL/L
(223.68 G/L, 3 N) AQUEOUS SOLUTION
Formula:
KCl
MW:
74.55 g/mol
Density:
1.13 g/cm³ (20°C)
Potassium Chloride Solution 3.0M
Made with deionized water.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
100 mL
Poly Bottle
Ea./
36.38
Cs. 12/
428.08
CONDUCTIVITY STANDARD 1,413 ΜS/
CM (25°C, POTASSIUM CHLORIDE 0.01
MOL/L AQUEOUS SOLUTION)
Formula:
KCl
MW:
74.55 g/mol
Density:
1 g/cm³ (20°C)
Potassium chloride solution 0.01M,
conductivity standard 1413 µmhos/cm.
Conductivity standard: 1413µS/m
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Shelf-life........................................................................................................................................... 1 year
Size
Packaging
Cat. No.
Price
1 L
Poly Bottle
Ea./
26.52
Cs. 12/
302.36
POTASSIUM CHROMATE
5% W/V AQUEOUS SOLUTION
Formula:
K
2
CrO
4
MW:
194.19 g/mol
CAS:
7789-00-6
EINECS:
232-140-5
Potassium Chromate (VI), Indicator Solution. 5% (w/v).
Made with deionized water.
Free from chlorine.................................................................................................................................Pass
For titration of chloride using silver nitrate......................................................................................... Suitable
Size Packaging Cat. No.
Price
500 mL Poly Bottle
Ea./
23.10
Cs. 12/
263.37
POTASSIUM FLUORIDE
50% AQUEOUS SOLUTION
Formula:
FK
MW:
58.1 g/mol
CAS:
7789-23-3
Potassium Fluoride, Solution. 50% (w/w).
Made with deionized water.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
pH..............................................................................................................................8.00-8.20 of 1:
6 dilution
Density........................................................................................................................................ 1.28-1.33
Size
Packaging
Cat. No.
Price
1 L
Poly Bottle
Ea./
70.54
Cs. 12/
804.20
CAS:
7447-40-7
EINECS:
231-211-8
CAS:
7447-40-7
EINECS:
231-211-8
ADR:
6.1,III
Index:
024-006-00-8
UN:
3287
EINECS:
232-151-5
ADR:
6.1,III
UN:
3422
POTASSIUM HYDROGEN PHTHALATE
Phthalic acid potassium salt, KHP
Formula:
C
8
H
5
KO
4
MW:
204.22 g/mol
Melting Pt:
295 to 300 °C
Density:
1.6362 g/cm³ (20°C)
CAS:
877-24-7
EINECS:
212-889-4
Potassium Hydrogen Phthalate, ACS
White solid
Assay....................................................................................................................99.95 – 100.05% C
8
H
5
O
4
K
pH of a 0.05M solution at 25.0 ± 0.2°C.......................................................................................... 4.00 – 4.02
Insoluble matter....................................................................................................................... 0.005% max
Chloride compounds (as Cl)........................................................................................................ 0.003% max
Sulfur compounds (as S)............................................................................................................ 0.002% max
Heavy metals (as Pb)................................................................................................................... 5 ppm max
Iron (Fe).................................................................................................................................... 5 ppm max
Sodium (Na)............................................................................................................................. 0.005% max
Size
Packaging
Cat. No.
Price
125 g
Poly Bottle
Ea./
47.58
Cs. 12/
436.86
500 g
Poly Bottle
Ea./
89.29
Cs. 12/
936.41
Increase regulatory
compliance with
VWR
CATALYST
Chemical
Tracking Services
VWR
CATALYST can make sure you
know the location and usage of every chemical at
your site. With industry-leading chemical tracking and
management technology, we provide the information
you need to comply with all federal, state, and local
regulations. For more information, connect with your
VWR Sales Representative or call
888.793.2300
.
O
O
OK
OH
46
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
P
Potassium Chloride
KCl
KF
KCl
K
2
CrO
4
1...,54,55,56,57,58,59,60,61,62,63 65,66,67,68,69,70,71,72,73,74,...100
Powered by FlippingBook